Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals
Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals
Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals
Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF043282.AP500 mL
Catalog number ALF043282.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 % (Coulometric)
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
Color scale=<10 APHA
Infrared spectrumConforms
GC>=98.5 %
View more
Trimethylamine solution used in research. As flavoring agents and adjuvants. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators, strongly basic anion exchange resins, dye leveling agents and a number of basic dyes.