Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 75-36-5 | C2H3ClO | 78.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Catalog number ALF043262.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS95-73-8
IUPAC Name2,4-dichloro-1-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyFUNUTBJJKQIVSY-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful an important reactant in Friedel-Crafts reactions as well as the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.