5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
Reagent for precipitation of proteins
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF043144.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3796-23-4
IUPAC Name3-(trifluoromethyl)pyridine
Molecular FormulaC6H4F3N
InChI KeyJTZSFNHHVULOGJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4150-1.4200 @ 20?C
Assay (GC)≥96.0%
Reagent for precipitation of proteins5-Sulfosalicylic acid dihydrate is used as a metal scavenger and it forms proton-transfer dye complexes with diazo compounds such as 4-(phenyldiazenyl)aniline. It is employed as a preparative measure for removing proteins prior to chromatographic analysis. It plays a vital role in reducing and fixation of proteins in agarose and polyacrylamide gels. It acts as a surfactant and lubricating grease additive. It is also used in urine tests to determine urine protein content. Further, it acts as an intermediate in the synthesis of active pharmaceutical ingredients like doxycycline and methacycline. In addition to this, it is used as a spray reagent for the detection of amino acids on thin layer chromatography (TLC) plates and as a doping agent for polyaniline.