Sulfuric acid, 3.0N Standardized Solution
Sulfuric acid, 3.0N Standardized Solution
Sulfuric acid, 3.0N Standardized Solution
Sulfuric acid, 3.0N Standardized Solution
Thermo Scientific Chemicals
Sulfuric acid, 3.0N Standardized Solution
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF043082.K21 L
Catalog number ALF043082.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Quantity1 L
Health Hazard 1H314-H335
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).