Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF043022.30250 g
Catalog number ALF043022.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
ColorSilver to Gray
CAS7429-90-5
Health Hazard 1H250-H261-H500
Health Hazard 3P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c