Sodium nitrite, 0.1M Standardized Solution
Sodium nitrite, 0.1M Standardized Solution
Sodium nitrite, 0.1M Standardized Solution
Sodium nitrite, 0.1M Standardized Solution
Thermo Scientific Chemicals
Sodium nitrite, 0.1M Standardized Solution
CAS: 7632-00-0 | NNaO2 | 69.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF043015.AP500 mL
Catalog number ALF043015.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS100-60-7
IUPAC NameN-methylcyclohexanamine
Molecular FormulaC7H15N
InChI KeyXTUVJUMINZSXGF-UHFFFAOYSA-N
SMILESCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow to brown
Infrared spectrumConforms
Refractive index1.4535 to 1.4575 (20°C, 589 nm)
Sodium nitrite, 0.1M standardized solution is used for volumetric analysis in laboratories.