Platinum wire, 0.3mm (0.012in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.3mm (0.012in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.3mm (0.012in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.3mm (0.012in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum wire, 0.3mm (0.012in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Form: Approx.1.5g/m, 0.477g/ft.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
25 cm
Catalog number ALF043014.BY
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 cm
Request bulk or custom format
Chemical Identifiers
CAS1665-00-5
IUPAC Namedichloro(²H₂)methane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-DICFDUPASA-N
SMILES[2H]C([2H])(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay by NMR>/= 99.8% (isotopic)