Multi-Element Plasma Standard Solution 4, Specpure™
Multi-Element Plasma Standard Solution 4, Specpure™
Thermo Scientific Chemicals
Multi-Element Plasma Standard Solution 4, Specpure™
Have Questions?
Catalog NumberQuantity
ALF042885.AE100 mL
Catalog number ALF042885.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS97-53-0
IUPAC Name2-methoxy-4-(prop-2-en-1-yl)phenol
Molecular FormulaC10H12O2
InChI KeyRRAFCDWBNXTKKO-UHFFFAOYSA-N
SMILESCOC1=CC(CC=C)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Specific gravity(20°C) 1.064 to 1.070
Appearance (Color)Clear colorless to yellow-green