Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Thermo Scientific Chemicals
Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
CAS: 3618-43-7 | C31H28N2Na4O13S
Have Questions?
Catalog NumberQuantity
ALF042773.AE100 mL
Catalog number ALF042773.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialXylenol Orange tetrasodium salt
CAS3618-43-7
Recommended StorageAmbient temperatures
EINECS Number222-805-8
TSCAYes