Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Thermo Scientific Chemicals
Methylene Blue, 1% w/v aq. soln.
CAS: 122965-43-9 | C16H18ClN3S
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF042771.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1949720-50-6
IUPAC Namedisodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC21H27N7Na2O14P2
InChI KeyQRGNQKGQENGQSE-MWLGWILCNA-L
SMILES[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<8.0 % (K.F.)
Appearance (Color)White to yellow
HPLC>=95.0 %
Infrared spectrumConforms
Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.