Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals
AA4272918
Have Questions?
Catalog NumberQuantity
ALF042729.A11 kg
Catalog number ALF042729.A1
Price (MYR)
1,720.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,720.00
Add to cart
Specifications
Chemical Name or MaterialSilica gel 60
Physical FormPowder
CAS7631-86-9
Health Hazard 1H373: May cause damage to organs through prolonged or repeated exposure.
Recommended StorageAmbient temperatures
View more
Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.