Silica gel 60, 0.062-0.105mm (150-230 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.062-0.105mm (150-230 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.062-0.105mm (150-230 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.062-0.105mm (150-230 mesh), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel 60, 0.062-0.105mm (150-230 mesh), Thermo Scientific Chemicals
Column chromatography without applied pressure (gravity pulled)
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALF042727.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilica gel 60
Physical Form-150+230 Mesh,S.A. 500-600m
CAS7631-86-9
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
View more
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.