Polystyrene latex microsphere, 0.50 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 0.50 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 0.50 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 0.50 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals
Polystyrene latex microsphere, 0.50 micron, 2.5 wt% dispersion in water
CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
ALF042714.4Y15 mL
Catalog number ALF042714.4Y
Price (MYR)
3,496.00
Add to cart
Quantity:
15 mL
Request bulk or custom format
Price (MYR)
3,496.00
Add to cart
Specifications
Chemical Name or MaterialPolystyrene latex microsphere
CAS9003-53-6
Physical FormDispersion
Storage Note 12°C to 8°C
Recommended StorageKeep cold
View more
Polystyrene latex microspheres are utilized in flow cytometry, fluorescence microscopy, and as calibration particles. It finds a use in the detection of trace amounts of antigens or antibodies in serum, urine, and cerebrospinal fluids. It is also employed as a stimulus responsive particulate emulsifier for oil-in-water emulsion.