Platinum Ruthenium foil, 0.05mm (0.002in) thick, Thermo Scientific Chemicals
Platinum Ruthenium foil, 0.05mm (0.002in) thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum Ruthenium foil, 0.05mm (0.002in) thick, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF042686.FI
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS14221-01-3
IUPAC Nametetrakis(triphenylphosphane) palladium
Molecular FormulaC72H60P4Pd
InChI KeyNFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
Carbon (C)73.7 to 76.0 %
Appearance (Form)Powder or crystals