Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALF042679.AP
Price (MYR)
299.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
299.00
Add to cart
Specifications
Chemical Name or MaterialPotassium chloride
Name Note1413μmho at 25°C
CAS7447-40-7
Quantity500 mL
CAS7732-18-5
View more
Potassium chloride 0.010M solution is used as conductive standard and as electrolyte in analytical chemistry.