Ammonium dihydrogen phosphate, Matrix Modifier Solution, Specpure™
Ammonium dihydrogen phosphate, Matrix Modifier Solution, Specpure™
Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, Matrix Modifier Solution, Specpure™
Have Questions?
Catalog NumberQuantity
ALF042663.AE100 mL
Catalog number ALF042663.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium dihydrogen phosphate Matrix Modifier Solution
Quantity100 mL
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390