Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals
Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals
Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals
Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals
CAS: 28210-41-5
Have Questions?
Catalog NumberQuantity
ALF042653.22100 g
Catalog number ALF042653.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly (styrene sulfonic acid)
Name NoteM.W. 75,000
CAS28210-41-5
Health Hazard 1H314-H335-H350
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more
Poly(styrene sulfonic acid) is used in water softening and to remove hard ions like calcium and magnesium. In the medical field, it is used as potassium binders in acute and chronic kidney disease. Its sodium salt is used as a superplastifier in cement, as a dye improving agent for cotton, as proton exchange membranes in fuel cell applications. It acts as a solid acid catalyst in organic synthesis. It is an antistatic resin used for electrographic and electrophotographic substrates.