Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
CAS: 115383-22-7 | C70 | 840.77 g/mol
Have Questions?
Catalog NumberdimQuantity
ALF042600.MD
Catalog number ALF042600.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Solubility InformationSoluble in benzene and toluene. Insoluble in water.
Unit Size
Fullerene powder is used for biomedical applications including the design of high-performance magnetic resonance imaging (MRI) contrast agents, X-ray imaging contrast agents, photodynamic therapy and drug and gene delivery. It is used in superconductors, photoactive polymers, film material modifiers, abrasive agent for cutting and grinding applications.