Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals
Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals
Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals
Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals
Base used in preparation of dienes and enolates
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF042595.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS99-50-3
IUPAC Name3,4-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyYQUVCSBJEUQKSH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to cream to brown
Assay (Silylated GC)≥96.0%
FormCrystals or powder or crystalline powder
Lithium bis(trimethylsilyl)amide is a base used in preparation of dienes and enolates. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. Lithium bis(trimethylsilyl)amide is also used in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only