ICP-MS Tuning Standard solution for HP∣r, Specpure™
ICP-MS Tuning Standard solution for HP∣r, Specpure™
Thermo Scientific Chemicals
ICP-MS Tuning Standard solution for HP∣r, Specpure™
Have Questions?
Catalog NumberQuantity
ALF042586.AE100 mL
Catalog number ALF042586.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS621-54-5
IUPAC Name3-(3-hydroxyphenyl)propanoic acid
Molecular FormulaC9H10O3
InChI KeyQVWAEZJXDYOKEH-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)110.0-116.0?C
Appearance (Color)Cream to brown