Sulfuric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Sulfuric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Sulfuric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Sulfuric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF042552.K21 L
Catalog number ALF042552.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS2893-78-9
IUPAC Namesodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Molecular FormulaC3Cl2N3NaO3
InChI KeyMSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals or granules
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dry substance)
Loss on drying=<5 %
Sulfuric acid has an useful applications in domestic acidic drain cleaner,as a drying agent, electrolyte in lead-acid batteries and various cleaning agents. It is also useful for mineral processing, fertilizer manufacturing, oil refining, waste water processing, and chemical synthesis. The sulfuric acid doped poly (m-aminophenol) is used as aliphatic alcohol vapor sensor material. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide, shows an useful application in sulfur-iodine cycle for hydrogen production.