Platinum foil, 0.05mm (0.002in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Platinum foil, 0.05mm (0.002in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Platinum foil, 0.05mm (0.002in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Platinum foil, 0.05mm (0.002in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Platinum foil, 0.05mm (0.002in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum foil, 0.05mm (0.002in) thick, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Form: approx. 0.67g/25x25mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF042456.FI
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS409-21-2
IUPAC Namemethanidylidynesilylium
Molecular FormulaCSi
InChI KeyHBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES[C-]#[Si+]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentHardness: 9 moh / 2500 knoop diamond
Assay from Supplier's CofA≥98.0% SiC
Impurity contentIron(III) oxide: ≤0.2%
Particle SizeAverage grain size 300-425µm by FEPA-Standard 42-1:2006
CommentAccepted on supplier's Certificate of Analysis