Magnesium, Oil based standard solution, Specpure™ Mg 5000μg/g
Magnesium, Oil based standard solution, Specpure™ Mg 5000μg/g
Magnesium, Oil based standard solution, Specpure™ Mg 5000μg/g
Magnesium, Oil based standard solution, Specpure™ Mg 5000μg/g
Thermo Scientific Chemicals
Magnesium, Oil based standard solution, Specpure™ Mg 5000μg/g
Have Questions?
Catalog NumberQuantity
ALF042443.1850 g
Catalog number ALF042443.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS530-93-8
IUPAC Name1,2,3,4-tetrahydronaphthalen-2-one
Molecular FormulaC10H10O
InChI KeyKCKZIWSINLBROE-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5585-1.5615 @ 20?C