Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Water <100ppm, typically 50ppm (0.005%)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF042311.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS87-69-4
IUPAC Name2,3-dihydroxybutanedioic acid
Molecular FormulaC4H6O6
InChI KeyFEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILESOC(C(O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or powder
Assay from Supplier's CofA≥98.5% (U.S. sourced material)
CommentMaterial Sourced in the U.S. and in other countries
Identification (FTIR)Conforms
Melting Point164-172°C (non-U.S. sourced material)
View more
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.