Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, anhydrous, 99.8+%, Thermo Scientific Chemicals
Water <100ppm, typically 50ppm (0.005%)
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number ALF042311.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS22059-21-8
IUPAC Name1-aminocyclopropane-1-carboxylic acid
Molecular FormulaC4H7NO2
InChI KeyPAJPWUMXBYXFCZ-UHFFFAOYSA-N
SMILESNC1(CC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.