Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals
Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals
Acrylic-DVB; Weakly basic gel type resin
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number ALF042253.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS824-46-4
IUPAC Name2-methoxybenzene-1,4-diol
Molecular FormulaC7H8O3
InChI KeyLAQYHRQFABOIFD-UHFFFAOYSA-N
SMILESCOC1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale yellow to brown or grey
Assay from Supplier's CofA≥96.0% (U.S. sourced material)
FormPowder
CommentMaterial sourced in the U.S. and in other countries
Assay (HPLC)≥96.0% (non-U.S. sourced material)
Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.