Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals
Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals
Acrylic-DVB; Weakly basic gel type resin
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number ALF042253.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS694-82-6
IUPAC Name2-fluorocyclohexan-1-one
Molecular FormulaC6H9FO
InChI KeyVQYOFTVCYSPHPG-UHFFFAOYNA-N
SMILESFC1CCCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4415-1.4475 @ 20?C
Appearance (Color)Clear colorless to brown or pink
Assay (GC)≥95.0%
FormLiquid
Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.