Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity, Thermo Scientific Chemicals
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity, Thermo Scientific Chemicals
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity, Thermo Scientific Chemicals
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity, Thermo Scientific Chemicals
Nafion∣r N-117 membrane, 0.180mm thick, ∣>0.90 meq/g exchange capacity, Thermo Scientific Chemicals
Widely used for proton exchange membrane (PEM) fuel cells and water electrolyzers. It is popular for separators in electrochemical cells requiring high ionic conductivity such as fuel cells and water electrolyzers.
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 30 cm
15 x 15 cm
Catalog number ALF042180.RF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
30 x 30 cm
Request bulk or custom format
Chemical Identifiers
CAS611-05-2
IUPAC Name3-methyl-4-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyXPAYEWBTLKOEDA-UHFFFAOYSA-N
SMILESCC1=CC(N)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale green
FormCrystals or powder or crystalline powder or flakes
Assay (HPLC)≥98.5%
Melting Point (clear melt)132.0-138.0?C
Nafion™ N-117 membrane is widely used for proton exchange membrane (PEM) fuel cells and water electrolyzers. It is popular for separators in electrochemical cells requiring high ionic conductivity such as fuel cells and water electrolyzers. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.