n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals
n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals
n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals
n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals
For Pesticide Residue, Environmental and GC Analysis
Have Questions?
Catalog NumberQuantity
ALF042100.K21 L
Catalog number ALF042100.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPlate : 50x50 mm
Appearance (Color)Black
Thickness~2 mm
Pesticide residue, environmental and GC analysis.n-Hexane is used in spectrophotometry and environmental testing. It is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.