Chloropentacarbonylrhenium(I), 98%
Chloropentacarbonylrhenium(I), 98%
Chloropentacarbonylrhenium(I), 98%
Chloropentacarbonylrhenium(I), 98%
Thermo Scientific Chemicals
Chloropentacarbonylrhenium(I), 98%
CAS: 14099-01-5 | C5ClO5Re | 361.707 g/mol
Have Questions?
Catalog NumberQuantity
ALF042096.031 g
Catalog number ALF042096.03
Price (MYR)
1,474.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,474.00
Add to cart
Specifications
Chemical Name or MaterialChloropentacarbonylrhenium(I)
CAS14099-01-5
Health Hazard 1H302+H312+H332-H315-H320-H335
Health Hazard 2GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Pentacarbonylchlororhenium(I) was used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands a well as to prepare [3]-ferrocenophane complexes. It has also been employed in preparation of a series of mixed-valence compounds of rhenium and iron. These compounds find application in the study of charge-transfer processes. 