Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 14405-43-7 | C15H21GaO6 | 367.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF042086.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS590-36-3
IUPAC Name2-methylpentan-2-ol
Molecular FormulaC6H14O
InChI KeyWFRBDWRZVBPBDO-UHFFFAOYSA-N
SMILESCCCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4085-1.4125 @ 20?C
It is used in research on Ga-containing materials. Gallium oxide thin films can be created with atomic layer epitaxy (ALE) by combining gallium acetylacetonate with either water or ozone as the precursor. It can also be used for the synthesis of high purity gallium nitride nano-wires and nano-needles at low temperatures.