2-Hydroxyacetophenone, 97+%
2-Hydroxyacetophenone, 97+%
2-Hydroxyacetophenone, 97+%
2-Hydroxyacetophenone, 97+%
Thermo Scientific Chemicals
2-Hydroxyacetophenone, 97+%
CAS: 582-24-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberQuantity
ALF042013.065 g
Catalog number ALF042013.06
Price (MYR)
572.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
572.00
Add to cart
Specifications
Chemical Name or Material2-Hydroxyacetophenone
CAS582-24-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2-Hydroxyacetophenone can be used to produce 2-hydroxy-1-phenyl-ethanone-(1)-oxime by heating. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only