Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Thermo Scientific Chemicals
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
ALF041937.031 g
Catalog number ALF041937.03
Price (MYR)
543.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
543.00
Add to cart
Specifications
Chemical Name or MaterialPolystyrene standard
Name NoteM.W. 290,000, Mw/Mn 1.06
CAS9003-53-6
Physical FormPowder