(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals
(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals
(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals
(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals
CAS: 112-02-7 | C19H42ClN | 320.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF041861.1425 g
Catalog number ALF041861.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5921-73-3
IUPAC Namenon-2-yn-1-ol
Molecular FormulaC9H16O
InChI KeyXERDOEKKHDALKJ-UHFFFAOYSA-N
SMILESCCCCCCC#CCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4530-1.4590 @ 20?C
Appearance (Color)Clear colourless to pale yellow
Assay (GC)≥94.0%
(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.