Chromium powder, APS <10 micron, 99.2% (metals basis), Thermo Scientific Chemicals
Chromium powder, APS &lt;10 micron, 99.2% (metals basis), Thermo Scientific Chemicals
Chromium powder, APS &lt;10 micron, 99.2% (metals basis), Thermo Scientific Chemicals
Chromium powder, APS &lt;10 micron, 99.2% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromium powder, APS <10 micron, 99.2% (metals basis), Thermo Scientific Chemicals
CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF041797.30250 g
Catalog number ALF041797.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS611-09-6
IUPAC Name5-nitro-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4N2O4
InChI KeyUNMYHYODJHKLOC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NC(=O)C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=96.0 %