Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF041748.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS939-97-9
IUPAC Name4-tert-butylbenzaldehyde
Molecular FormulaC11H14O
InChI KeyOTXINXDGSUFPNU-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=94 %
Refractive index1.5270 to 1.5290 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Humic acid sodium salt is a principal component of humic substances. It boosts plant growth.