Humic acid, Thermo Scientific Chemicals
Humic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid, Thermo Scientific Chemicals
CAS: 1415-93-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF041747.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2516-33-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4290 to 1.4330 (20°C, 589 nm)
Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.