Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
CAS: 2695-37-6 | C8H7NaO3S | 206.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ALF041690.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialStyrene-4-sulfonic acid sodium salt
CAS2695-37-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Styrene-4-sulfonic acid sodium salt is grafted on poly(ethersulfone) membrane to determine the fractionation of proteins with similar molecular weights. It is used as sodium based ion exchange membranes for membrane capacitive deionization (MCDI) for synthesis. It is used in the study of copolymerization characteristics of sodium styrenesulfonate.