Silver powder, spherical, -500 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Silver powder, spherical, -500 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Silver powder, spherical, -500 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Silver powder, spherical, -500 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver powder, spherical, -500 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF041602.0910 g
Catalog number ALF041602.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS79-44-7
IUPAC NameN,N-dimethylcarbamoyl chloride
Molecular FormulaC3H6ClNO
InChI KeyYIIMEMSDCNDGTB-UHFFFAOYSA-N
SMILESCN(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4500-1.4560 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%