Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals
Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals
Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals
Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals
Useful chiral synthon; building block for depsipeptides
Have Questions?
Catalog NumberQuantity
ALF041529.AK250 mL
Catalog number ALF041529.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS145224-94-8
IUPAC Name4-(2-sulfonatoethyl)morpholin-4-ium
Molecular FormulaC6H13NO4S
InChI KeySXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CC[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (as monohydrate)
Identification (FTIR)Conforms
Appearance (Color)White
Useful chiral synthon; building block for depsipeptides.Sodium DL-lactate, 60% w/w aq. Soln is used as a food additive, plasticizer for casein, preservative, acidity regulator, bulking agent, and in shampoo and liquid soaps. It is also used for the treatment for arrhythmias and sympathomimetics. It is useful chiral synthon and building block for depsipeptides. It is used as a corrosion inhibitor in alcohol antifreeze mixtures, flavoring agent, emulsifier, corrosion inhibitor and pH control agent.