Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF041496.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight571.47
Unit Size
Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.