Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number ALF041467.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS244193-56-4
IUPAC Name1-decyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular FormulaC14H27BF4N2
InChI KeyQGUMDWFYJYXDTH-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCCCCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>96.0%
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.