Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals
Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals
Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals
Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals
CAS: 18472-51-0 | C34H54Cl2N10O14 | 897.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
4 x 500 mL
Catalog number ALF041385.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS127-68-4
IUPAC Namesodium 3-nitrobenzene-1-sulfonate
Molecular FormulaC6H4NNaO5S
InChI KeyLJRGBERXYNQPJI-UHFFFAOYSA-M
SMILES[Na+].[O-][N+](=O)C1=CC=CC(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Appearance (Color)White to yellow
HPLC>=96.0 %
Infrared spectrumConforms
Water=<3.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorhexidine digluconate, 20% w/v aq. Soln is used as an oral antiseptic agent and anti-microbial agent. It is an active ingredient in germicidal mouthwash, pet shampoo, surgical lubricants and wipes and hand soap. It is utilized in the study to know the role of essential oils for improving skin antisepsis. It plays a vital role as disinfectants and preservative in pharmaceutical products. It is also used as an additive in cosmetics such as creams, toothpaste and deodorants.
Solubility
Miscible with water, ethanol and acetone.
Notes
Store in a cool place. Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Rabe, P.; Twetman, S.; Kinnby, B.; Svensäter, G.; Davies, J. R. Effect of Fluoride and Chlorhexidine Digluconate Mouthrinses on Plaque Biofilms. Open Dent. J. 2015, 9, 106-111.
  2. Bueno, M. D. F. T.; Basting, R. T.; Turssi, C. P.; França, F. M. G.; Amaral, F. L. B. Effect of 2% chlorhexidine digluconate application and water storage on the bond strength to superficial and deep dentin. J. Adhes. Sci. Technol. 2015, 29 (12), 1258-1267.
  3. Vadhana, S.; Latha, J.; Velmurugan, N. Evaluation of penetration depth of 2% chlorhexidine digluconate into root dentinal tubules using confocal laser scanning microscope. Restor. Dent. Endod. 2015, 40 (2), 149-154.