Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis), Thermo Scientific Chemicals
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis), Thermo Scientific Chemicals
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis), Thermo Scientific Chemicals
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lead granules, 0.6-4.0mm, (0.02-0.16in), 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Catalog NumberQuantity
ALF041370.22100 g
Catalog number ALF041370.22
Price (MYR)
2,016.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,016.00
Add to cart
Specifications
Chemical Name or MaterialLead
ColorGray
CAS7439-92-1
Health Hazard 1H350-H360FD-H362-H372
Health Hazard 3P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c