Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
CAS: 12449-35-3 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF040216.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2896-60-8
IUPAC Name4-ethylbenzene-1,3-diol
Molecular FormulaC8H10O2
InChI KeyVGMJYYDKPUPTID-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)94.0-100.0?C
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.