Sodium tetraborate decahydrate, ACS, 99.5-105.0%
Sodium tetraborate decahydrate, ACS, 99.5-105.0%
Sodium tetraborate decahydrate, ACS, 99.5-105.0%
Sodium tetraborate decahydrate, ACS, 99.5-105.0%
Thermo Scientific Chemicals
Sodium tetraborate decahydrate, ACS, 99.5-105.0%
CAS: 1303-96-4 | B4Na2O7·10H2O
Have Questions?
Catalog NumberQuantity
ALF040114.36500 g
Catalog number ALF040114.36
Price (MYR)
362.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
362.00
Add to cart
Specifications
Chemical Name or MaterialSodium tetraborate decahydrate
Melting Point75°C
CAS1303-96-4
ColorGray
Health Hazard 1H319-H360FD
View more
It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.