Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol
Have Questions?
Catalog NumberQuantity
ALF039781.K21 L
Catalog number ALF039781.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid disodium salt
CAS139-33-3
ColorColorless
Recommended StorageAmbient temperatures
EINECS Number205-358-3
View more
It is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.