Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
CAS: 115383-22-7 | C70 | 840.77 g/mol
Have Questions?
Catalog NumberQuantity
ALF039720.MF50 mg
Catalog number ALF039720.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS58236-70-7
IUPAC Name5-bromo-3-chloro-1,2-dihydropyridin-2-one
Molecular FormulaC5H3BrClNO
InChI KeyIPWYUROVLMVBGD-UHFFFAOYSA-N
SMILESClC1=CC(Br)=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.