Vanadium(IV) etioporphyrin III oxide
Vanadium(IV) etioporphyrin III oxide
Vanadium(IV) etioporphyrin III oxide
Vanadium(IV) etioporphyrin III oxide
Thermo Scientific Chemicals
Vanadium(IV) etioporphyrin III oxide
CAS: 15709-03-2 | C32H38Cl2N4OV | 616.53 g/mol
Have Questions?
Catalog NumberQuantity
ALF039586.MF50 mg
Catalog number ALF039586.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS42712-64-1
IUPAC Name2-amino-1,4-dihydroquinolin-4-one
Molecular FormulaC9H8N2O
InChI KeyLWGUCIXHBVVATR-UHFFFAOYSA-N
SMILESNC1=CC(=O)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤12%
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (dry wt. basis)
Appearance (Color)White to pale brown
Vanadium(IV) etioporphyrin III oxide is used to check the trapping efficiency of the external AlOxlayer in AlOxcoating of ultrastable Zeolite.