Diethylzinc, 96%
Diethylzinc, 96%
Diethylzinc, 96%
Thermo Scientific Chemicals
Diethylzinc, 96%
CAS: 557-20-0 | C4H10Zn | 123.50 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALF039564.22100 gCylinder
Catalog number ALF039564.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Cylinder
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylzinc
CAS557-20-0
Health Hazard 1H225-H250-H260-H314-H335
Health Hazard 2GHS H Statement
H250-H260-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
View more
It is used as an important reagent in organic chemistry. It is used in organic synthesis as a source of the ethyl carbanion in addition reactions to carbonyl groups.